Product Details

SNP ID
rs6187389
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:86170027 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGATGACAGATGCTGTGGCCCACA[A/G]CCCAAAAGGACCAAGCCCACACCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cerk PubMed Links

Gene Details

Gene
Cerk
Gene Name
ceramide kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145475.4 Intron NP_663450.3

View Full Product Details