Product Details

SNP ID
rs13476969
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:9691409 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GACTCTGGCGGCCATGGCTGACAAA[C/T]TCAAACTGACAAAAGGAGACATCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pag1 PubMed Links

Gene Details

Gene
Pag1
Gene Name
phosphoprotein associated with glycosphingolipid microdomains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195031.1 4267 UTR 3 NP_001181960.1
NM_053182.5 4267 UTR 3 NP_444412.2

View Full Product Details