Product Details

SNP ID
rs3680295
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:188900889 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTGGACCAGCTCCACCCAACT[T/C]AGATTATTCAGAGTCCAGCAGAATC
Phenotype
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
Ush2a PubMed Links

Gene Details

Gene
Ush2a
Gene Name
Usher syndrome 2A (autosomal recessive, mild)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021408.3 Intron NP_067383.3

View Full Product Details