Product Details

SNP ID
rs3658142
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:29813265 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGTTACAACACACAAGACCCTGTCT[C/T]AGCAACAACAAAGCAGCAAGAAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Slc27a4 PubMed Links

Gene Details

Gene
Slc27a4
Gene Name
solute carrier family 27 (fatty acid transporter), member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011989.4 Intron NP_036119.1

View Full Product Details