Product Details
- SNP ID
-
rs3022940
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:147362411 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCGGTGGAGATAACCAAGCCGCA[G/A]CCCGGAAAGTTGAAGAGACACTTTG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AI646519
PubMed Links
Gene Details
- Gene
- AI646519
- Gene Name
- expressed sequence AI646519
There are no transcripts associated with this gene.
- Gene
- Pax1
- Gene Name
- paired box 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008780.2 |
|
Intron |
|
|
NP_032806.2 |
View Full Product Details