Product Details

SNP ID
rs3023233
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:28596047 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGGACTTGATCAGTTTCTGTGACT[A/G]TATGAGAGCATCCACATTTCATGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ptprk PubMed Links

Gene Details

Gene
Ptprk
Gene Name
protein tyrosine phosphatase, receptor type, K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008983.2 4827 UTR 3 NP_033009.1

View Full Product Details