Product Details

SNP ID
rs3663879
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:82863758 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAACAGAGAAACATGAACACC[G/C]GGCACTCCTGAACAGGGCCACCTGT
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
Rad51d PubMed Links

Gene Details

Gene
Rad51d
Gene Name
RAD51 homolog D
There are no transcripts associated with this gene.

Gene
Rffl
Gene Name
ring finger and FYVE like domain containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007465.3 Intron NP_001007466.1
NM_001164569.1 Intron NP_001158041.1
NM_001164570.1 Intron NP_001158042.1
NM_001164571.1 Intron NP_001158043.1
NM_026097.3 Intron NP_080373.1

View Full Product Details