Product Details
- SNP ID
-
rs3677240
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44163125 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACCACCTTGTTCCTAAACTATGT[G/A]AAGAGGTCTTATATAAATTCCCCAG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Clic5
PubMed Links
Gene Details
- Gene
- Clic5
- Gene Name
- chloride intracellular channel 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172621.2 |
|
Intron |
|
|
NP_766209.1 |
View Full Product Details