Product Details

SNP ID
rs3681920
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:25660288 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGGATCTCAGTGCCATAGGGA[C/T]AGAAATGCGCTGCTCAGTCCTCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Celf4 PubMed Links

Gene Details

Gene
Celf4
Gene Name
CUGBP, Elav-like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146292.1 Intron NP_001139764.1
NM_001146293.1 Intron NP_001139765.1
NM_001146294.1 Intron NP_001139766.1
NM_001146295.1 Intron NP_001139767.1
NM_001174074.1 Intron NP_001167545.1
NM_133195.3 Intron NP_573458.2

View Full Product Details