Product Details

SNP ID
rs3699591
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.X:49792536 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCAACACTTGCAGCACAAGGT[T/C]GAAGGAGGTCAATAGGGAGATAGCT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Igsf1 PubMed Links

Gene Details

Gene
Igsf1
Gene Name
immunoglobulin superfamily, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177591.4 Intron NP_808259.2
NM_177915.4 Intron NP_808583.1
NM_183335.2 Intron NP_899178.2
NM_183336.2 Intron NP_899179.1

View Full Product Details