Product Details
- SNP ID
-
rs13475825
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:40312894 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGACATAAAAAGGAGTCTGGGA[A/T]GAGCATCTTTGAGTGTTGAATTCAG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Il1r1
PubMed Links
Gene Details
- Gene
- Il1r1
- Gene Name
- interleukin 1 receptor, type I
View Full Product Details