Product Details

SNP ID
rs4224578
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:89306404 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTATTTTGCATAGAGTAACCTG[C/T]TACCACTACAATTCATCACTGGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cdkn2b PubMed Links

Gene Details

Gene
Cdkn2b
Gene Name
cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007670.4 1278 UTR 3 NP_031696.1

View Full Product Details