Product Details

SNP ID
rs4224498
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:56744668 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACACCCCCCAGGTCAATGTGCTGCC[C/T]GAGAGAGACACTGCAGTCTGGACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Actl7a PubMed Links

Gene Details

Gene
Actl7a
Gene Name
actin-like 7a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009611.3 1247 Silent Mutation NP_033741.1
Gene
Actl7b
Gene Name
actin-like 7b
There are no transcripts associated with this gene.

Gene
Ikbkap
Gene Name
inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein
There are no transcripts associated with this gene.

View Full Product Details