Product Details

SNP ID
rs3676304
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:23525900 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGTCTGATTCAGCAAGTCTACAAT[A/G]GGACAGGATAACCAGCATTTTACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cadps2 PubMed Links

Gene Details

Gene
Cadps2
Gene Name
Ca2+-dependent activator protein for secretion 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252105.1 Intron NP_001239034.1
NM_001252106.1 Intron NP_001239035.1
NM_001252107.1 Intron NP_001239036.1
NM_001252108.1 Intron NP_001239037.1
NM_001252109.1 Intron NP_001239038.1
NM_001252110.1 Intron NP_001239039.1
NM_153163.4 Intron NP_694803.3

View Full Product Details