Product Details

SNP ID
rs4230267
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:34100272 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGGTCACATAGCTCTGCCAAT[C/T]GGGACACTCACTTAACGGAAATTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A630023A22Rik PubMed Links

Gene Details

Gene
A630023A22Rik
Gene Name
RIKEN cDNA A630023A22 gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251843.1 1505 Intron NP_001238772.1
Gene
Anxa8
Gene Name
annexin A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281845.1 1505 UTR 3 NP_001268774.1
NM_013473.4 1505 UTR 3 NP_038501.2

View Full Product Details