Product Details

SNP ID
rs13482770
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:24992136 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCTAAACCTATCTCCTGGGAGCAG[A/G]CACTTCCTCCTAGCTAGCCACCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Lpp PubMed Links

Gene Details

Gene
Lpp
Gene Name
LIM domain containing preferred translocation partner in lipoma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145952.1 14414 UTR 3 NP_001139424.1
NM_001145954.1 14414 UTR 3 NP_001139426.1
NM_178665.5 14414 UTR 3 NP_848780.3

View Full Product Details