Product Details

SNP ID
rs4222668
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:136921821 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTTACACCGTACTGGGTTGGGCTC[C/T]CTGCCCTCTTAATATTGCAGAACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Nr5a2 PubMed Links

Gene Details

Gene
Nr5a2
Gene Name
nuclear receptor subfamily 5, group A, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159769.2 Intron NP_001153241.1
NM_030676.3 Intron NP_109601.1

View Full Product Details