Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYRK2
Entrez Gene ID
102126248
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
Chromosome Location
Chr.11: 66653168 - 66744825 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571477.2 XP_005571534.1 1 - 2 530 74

View Full Product Details