Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLX1
Entrez Gene ID
102134887
Gene Name
T cell leukemia homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 100079079 - 100088248 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566234.2 XP_005566291.1 1 - 2 3507 61

View Full Product Details