Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNG12
Entrez Gene ID
102126944
Gene Name
G protein subunit gamma 12
Gene Aliases
-
Chromosome Location
Chr.1: 159500306 - 159638494 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543069.2 XP_005543126.1 1 - 2 429 103
XM_005543071.2 XP_005543128.1 1 - 2 429 103

View Full Product Details