Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CISD1
Entrez Gene ID
102141257
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
-
Chromosome Location
Chr.9: 77864883 - 77884262 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565815.2 XP_005565872.1 2 - 3 478 91
XM_005565816.2 XP_005565873.1 2 - 3 219 91
XM_015456349.1 XP_015311835.1 2 - 3 293 91

View Full Product Details