Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB4
Entrez Gene ID
102126189
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
-
Chromosome Location
Chr.16: 7519989 - 7545246 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582756.2 XP_005582813.1 2 - 3 325 154
XM_015437584.1 XP_015293070.1 2 - 3 226 154

View Full Product Details