Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD7
Entrez Gene ID
102135893
Gene Name
SMAD family member 7
Gene Aliases
-
Chromosome Location
Chr.18: 32862254 - 32893283 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586745.1 XP_005586802.1 3 - 4 951 105
XM_005586746.1 XP_005586803.1 3 - 4 948 105

View Full Product Details