Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102126225
Entrez Gene ID
102126225
Gene Name
developmental pluripotency-associated protein 4-like
Gene Aliases
-
Chromosome Location
Chr.3: 15539 - 16369 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015446681.1 XP_015302167.1 1 - 1 388 73

Gene Symbol
DPPA4
Entrez Gene ID
102116348
Gene Name
developmental pluripotency associated 4
Gene Aliases
-
Chromosome Location
Chr.2: 162835306 - 162856041 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015445460.1 XP_015300946.1 3 - 4 421 73

View Full Product Details