Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDH17
Entrez Gene ID
102131587
Gene Name
protocadherin 17
Gene Aliases
-
Chromosome Location
Chr.17: 37130762 - 37228640 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585937.2 XP_005585994.1 2 - 3 5208 58
XM_015439302.1 XP_015294788.1 2 - 3 5220 58
XM_015439303.1 XP_015294789.1 3 - 4 5115 58

View Full Product Details