Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A12
Entrez Gene ID
102115752
Gene Name
solute carrier family 2 member 12
Gene Aliases
GLUT-12, GLUT12
Chromosome Location
Chr.4: 38228347 - 38295078 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284710.1 NP_001271639.1 2 - 3 1612 90
GenBank mRNA AB056798.1 - 2 - 3 1612 90

View Full Product Details