Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
102116044
Gene Name
POU class 2 homeobox associating factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 107320231 - 107345897 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579572.2 XP_005579629.1 2 - 3 635 119

View Full Product Details