Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL16
Entrez Gene ID
102130695
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
Chromosome Location
Chr.16: 79308664 - 79311483 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585250.2 XP_005585307.1 4 - 5 423 81

View Full Product Details