Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102145815
Entrez Gene ID
102145815
Gene Name
glycine N-acyltransferase-like protein 1
Gene Aliases
GLYATL1
Chromosome Location
Chr.14: 15003518 - 15020364 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015434683.1 XP_015290169.1 2 - 3 947 94

View Full Product Details