Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEAL9
Entrez Gene ID
102120229
Gene Name
transcription elongation factor A like 9
Gene Aliases
-
Chromosome Location
Chr.X: 100182894 - 100184912 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594225.2 XP_005594282.1 2 - 3 204 116

View Full Product Details