Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIQK
Entrez Gene ID
102120817
Gene Name
triple QxxK/R motif containing
Gene Aliases
-
Chromosome Location
Chr.8: 94025401 - 94095193 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563683.2 XP_005563740.1 1 - 2 135 104
XM_005563684.2 XP_005563741.1 1 - 2 135 104

View Full Product Details