Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIPR1
Entrez Gene ID
101925116
Gene Name
EARP complex and GARP complex interacting protein 1
Gene Aliases
TSSC1
Chromosome Location
Chr.13: 106381252 - 106405569 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015434034.1 XP_015289520.1 4 - 5 856 88
GenBank mRNA AB169673.1 - - 1131 88
AB174551.1 - - 1010 88

View Full Product Details