Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP7
Entrez Gene ID
102135289
Gene Name
GTPase, IMAP family member 7
Gene Aliases
-
Chromosome Location
Chr.3: 183200178 - 183210296 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551144.2 XP_005551201.2 1 - 2 115 82
XM_015448278.1 XP_015303764.1 1 - 2 115 82
XM_015448279.1 XP_015303765.1 1 - 2 115 82

View Full Product Details