Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX12
Entrez Gene ID
102133445
Gene Name
testis expressed 12
Gene Aliases
-
Chromosome Location
Chr.14: 108153273 - 108158561 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579631.2 XP_005579688.1 1 - 2 64 67

View Full Product Details