Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLMP
Entrez Gene ID
102115806
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1H1orf85
Chromosome Location
Chr.1: 95427061 - 95430049 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541456.2 XP_005541513.1 5 - 6 1094 98

View Full Product Details