Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD49
Entrez Gene ID
102137143
Gene Name
ankyrin repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.14: 90342600 - 90348494 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579380.2 XP_005579437.1 2 - 3 442 152
XM_005579381.2 XP_005579438.1 2 - 3 457 152
XM_005579382.2 XP_005579439.1 2 - 3 370 152

View Full Product Details