Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL9
Entrez Gene ID
102141538
Gene Name
actin like 9
Gene Aliases
-
Chromosome Location
Chr.19: 9081543 - 9082975 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587851.2 XP_005587908.1 1 - 1 166 177

View Full Product Details