Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX53
Entrez Gene ID
102143520
Gene Name
DEAD-box helicase 53
Gene Aliases
-
Chromosome Location
Chr.X: 20941947 - 20944073 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593169.2 XP_005593226.1 1 - 1 1946 150

View Full Product Details