Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP7A1
Entrez Gene ID
102132452
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
-
Chromosome Location
Chr.8: 59604310 - 59617918 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563370.2 XP_005563427.1 1 - 2 217 96
XM_015454879.1 XP_015310365.1 2 - 3 142 96

View Full Product Details