Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIAH3
Entrez Gene ID
102131190
Gene Name
siah E3 ubiquitin protein ligase family member 3
Gene Aliases
-
Chromosome Location
Chr.17: 24974207 - 25046267 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585797.2 XP_005585854.1 1 - 2 274 98

View Full Product Details