Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTF1
Entrez Gene ID
102130856
Gene Name
cardiotrophin 1
Gene Aliases
-
Chromosome Location
Chr.20: 29541746 - 29550046 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591688.2 XP_005591745.1 2 - 3 267 78
XM_005591689.2 XP_005591746.1 2 - 3 223 78
XM_005591690.2 XP_005591747.1 2 - 3 204 78

View Full Product Details