Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC1A2
Entrez Gene ID
101866847
Gene Name
solute carrier family 1 member 2
Gene Aliases
EAAT2
Chromosome Location
Chr.14: 32016478 - 32180736 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578188.2 XP_005578245.1 1 - 2 617 67

View Full Product Details