Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83A
Entrez Gene ID
102136809
Gene Name
family with sequence similarity 83 member A
Gene Aliases
-
Chromosome Location
Chr.8: 124528220 - 124561123 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564019.2 XP_005564076.1 2 - 3 646 139
XM_015455147.1 XP_015310633.1 3 - 4 2702 139

View Full Product Details