Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDRT1
Entrez Gene ID
102139554
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
FBXW10
Chromosome Location
Chr.16: 15778849 - 15821921 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582955.2 XP_005583012.1 8 - 9 2315 125
XM_015437659.1 XP_015293145.1 9 - 10 2515 125
XM_015437660.1 XP_015293146.1 9 - 10 2515 125
XM_015437662.1 XP_015293148.1 9 - 10 1918 125
GenBank mRNA AB178997.1 - 4 - 5 466 125

View Full Product Details