Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYDGF
Entrez Gene ID
102120573
Gene Name
myeloid derived growth factor
Gene Aliases
C19H19orf10
Chromosome Location
Chr.19: 4743281 - 4754446 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587582.2 XP_005587639.1 5 - 6 552 134

View Full Product Details