Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLTRN
Entrez Gene ID
102131240
Gene Name
collectrin, amino acid transport regulator
Gene Aliases
TMEM27
Chromosome Location
Chr.X: 13451882 - 13489962 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593038.1 XP_005593095.1 1 - 2 305 139

View Full Product Details