Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102130310
Entrez Gene ID
102130310
Gene Name
IQ domain-containing protein F1
Gene Aliases
IQCF1
Chromosome Location
Chr.2: 107150186 - 107155700 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005547278.2 XP_005547335.1 1 - 2 108 117

View Full Product Details