Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA3
Entrez Gene ID
102132341
Gene Name
sperm acrosome associated 3
Gene Aliases
-
Chromosome Location
Chr.16: 29348830 - 29374752 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583414.2 XP_005583471.1 2 - 3 448 130
XM_015438161.1 XP_015293647.1 3 - 4 542 130
XM_015438163.1 XP_015293649.1 2 - 3 389 130

View Full Product Details