Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESPNL
Entrez Gene ID
102135315
Gene Name
espin like
Gene Aliases
-
Chromosome Location
Chr.12: 127943372 - 127973585 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574773.2 XP_005574830.1 1 - 2 1170 156

View Full Product Details