Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGLAP
Entrez Gene ID
102121548
Gene Name
bone gamma-carboxyglutamate protein
Gene Aliases
BGP
Chromosome Location
Chr.1: 95482470 - 95485022 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541474.2 XP_005541531.1 1 - 2 1530 105

View Full Product Details